Order Entry
Belgium
Orders LinkContactUsLinkComponent
Verso™ qRT-PCR Kits

Verso™ qRT-PCR Kits

Verso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 400×25 µl reactions, 1 KIT
Catalog # 733-1134
Verso™ qRT-PCR Kits
Catalog # 733-1134
CAS Number:  
Verso™ qRT-PCR Kits
Verso™ qRT-PCR Kits

Verso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 400×25 µl reactions, 1 KIT

CAS Number:  
Price
2 080,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

The Verso™ qRT-PCR system combines the Verso™ RT enzyme with ABsolute™ Blue qPCR master mix to provide maximum convenience and versatility. Available in a 1- or 2-step format, as well as multiple pack sizes, to suit all application and throughput requirements.

  • The combining of Verso™ enzyme and Thermo-Start™ Taq polymerase gives sensitive results from low copy number RNA template
  • Inert blue dye for easy visualisation of the qRT-PCR reaction
  • No need to perform an RNase H digestion after cDNA synthesis, therefore streamlining the reaction protocol
  • RT enhancer saves time by eliminating the need for DNase treatment of samples
  • Multiple priming options for optimisation of specific RNA templates
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Verso™ SYBR® Green 1-Step qRT-PCR ROX Kit, 400×25 µl reactions
  • Cat. No.:
    733-1134
  • One step:
    Yes
  • Application:
    RT-qPCR dye

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.