Verso™ qRT-PCR Kits
Verso™ 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactions, 1 KIT
733-1112
The Verso™ qRT-PCR system combines the Verso™ RT enzyme with ABsolute™ Blue qPCR master mix to provide maximum convenience and versatility. Available in a 1- or 2-step format, as well as multiple pack sizes, to suit all application and throughput requirements.
- The combining of Verso™ enzyme and Thermo-Start™ Taq polymerase gives sensitive results from low copy number RNA template
- Inert blue dye for easy visualisation of the qRT-PCR reaction
- No need to perform an RNase H digestion after cDNA synthesis, therefore streamlining the reaction protocol
- RT enhancer saves time by eliminating the need for DNase treatment of samples
- Multiple priming options for optimisation of specific RNA templates
- Pk:1 KIT
- Description:Verso™ 1-Step qRT-PCR Kit plus ROX vial, 200×25 µl reactions
- Cat. No.:733-1112
- One step:Yes
- Application:RT-qPCR dye