Order Entry
Belgium
Orders LinkContactUsLinkComponent
Round canisters

Round canisters

Blue liter scale, 10 L, Natural, S65, With handle and
measuring scale, 205×205×430 mm
Catalog # 590-0572
Supplier:   S.C.A.T.
Round canisters
Catalog # 590-0572
Supplier:  S.C.A.T.
Round canisters
Round canisters

Blue liter scale, 10 L, Natural, S65, With handle and
measuring scale, 205×205×430 mm

Price
96,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.

  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
  • Available in the common threads S65, S70/71 and B83
  • Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
  • Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances

SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.

Specifications
  • Thread:
    S65
  • Type:
    With handle and
    measuring scale
  • Capacity:
    10 L
  • Colour:
    Natural
  • Description:
    Blue liter scale
  • Seq:
    04
  • W×D×H:
    205×205×430 mm
  • Cat. No.:
    590-0572