ContactUsLinkComponent
Universal Plan Super Apochromat objective with 20x magnification. With working distance of 0.6 mm, numerical aperture of 0.75 and cover correction of 0.17 mm. 1 * 1 items
Catalog # OLYMN1480500
Supplier:  OLYMPUS OPTICAL
Universal Plan Super Apochromat objective with 20x magnification. With working distance of 0.6 mm, numerical aperture of 0.75 and cover correction of 0.17 mm. 1 * 1 items
Catalog # OLYMN1480500
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together