Orders LinkContactUsLinkComponent
Objectives for CKX53
Objectives for CKX53
LC achromatic objective with 1 mm correction and relief contrast (RC3), 1,9 mm
Catalog # OLYMN1478100
Objectives for CKX53
Catalog # OLYMN1478100
Supplier:  
Objectives for CKX53
Objectives for CKX53
LC achromatic objective with 1 mm correction and relief contrast (RC3), 1,9 mm
Discontinued
Product Details & Documents
Specifications
  • Magnification:
    40×/0,55
  • Description:
    LC achromatic objective with 1 mm correction and relief contrast (RC3)
  • Working distance:
    1,9 mm