Orders LinkContactUsLinkComponent
Cell viability assay kits
Cell viability assay kits
XTT Cell Viability Assay kit, 1000 assays, 1 KIT
Catalog # 30007.
Supplier:   Biotium
Cell viability assay kits
Catalog # 30007.
Supplier:  Biotium
CAS Number:  
Cell viability assay kits
XTT Cell Viability Assay kit, 1000 assays, 1 KIT
CAS Number:  
Price
308,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assaying cell viability and quantitating cell number. MTT and XTT are reduced to coloured formazin salts that can be measured by absorbance. MTT generates an insoluble formazin salt, requiring cell lysis before the absorbance can be measured, while XTT does not require cell lysis for measurement. Resazurin is a non-fluorescent blue dye that is reduced to the pink fluorescent compound resorufin, which can be measured by fluorescence or absorbance.
Documents
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    XTT Cell Viability Assay kit, 1000 assays
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.