Order Entry
Belgium
Orders LinkContactUsLinkComponent
Injection needles

Injection needles

Injection needles, Luer lock, regular, 15, 10 mm, 1,8 mm, Bevelled
Catalog # SOCO371.1810
Supplier:   Socorex Isba
Injection needles
Catalog # SOCO371.1810
Supplier:  Socorex Isba
Injection needles
Injection needles

Injection needles, Luer lock, regular, 15, 10 mm, 1,8 mm, Bevelled

Price
30,60 €
Log in for availability
1 * 12 ST
Product Details & Documents

Reusable Luer lock needles intended for safe, precise animal injections. Sharp bevel and made of stainless steel for long durability.

Delivery information: Supplied in dispensing box with sliding lid for easy and safe needle removal.

Specifications
  • Ø:
    1,8 mm
  • Gauge:
    15
  • Type of tip:
    Bevelled
  • Description:
    Injection needles, Luer lock, regular
  • Length:
    10 mm
  • Seq:
    02