Redox electrodes, InLab® Redox
InLab® Redox, S7, Ceramic, 3 mol/l KCl, Glass body, platinum ring, 0...+100 °C, 12×120 mm
Catalogus # 662-2877
Leverancier: Mettler - Toledo
Ga naar:
Product Details & Documents
Redox electrodes with S7 connector.
- Ideal for testing the oxidation/reduction in factory and industrial waste (chromium and cyanide concentration), fish farming and aquariums (water quality), and swimming pools (sanitation levels)
- ARGENTHAL™ reference system with silver-ion trap (InLab® Redox Micro: without silver-ion trap)
- EQUITHAL® technology with symmetrical conducting layers guarantees shortest response times despite large changes in temperature
- Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
Temperature range: 0 to 100 °C
pH range: 0 - 14
Temperature range: 0...+100 °C
Ordering information: Supplied with test certificate and without cable. Other electrodes on request.
Documents
Mettler InLab® Sensors
Product Literature
Specificaties
- Connection:S7
- Ø×L:12×120 mm
- Description:InLab® Redox
- Material:Glass body, platinum ring
- Seq:01
- Diaphr.:Ceramic
- Electrolyte:3 mol/l KCl
- Temp.:0...+100 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



