pH electrodes, InLab®, Expert Series
InLab® Expert NTC30, InLab® Expert NTC30, 0 - 14, 0...+100 °C, PEEK, XEROLYT® polymer, 12×120 mm, MultiPin™, Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samples
Catalog # 662-2843
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
- ARGENTHAL™ reference system with silver-ion trap
- Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
- Unbreakable PEEK shaft
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 100 °C
Temperature probe: NTC30 kOhm for InLab® Expert Pro and InLab® Expert NTC30; PT1000 for InLab® Expert DIN and InLab® Expert PT1000
Ordering information: Supplied with test certificate.
Documents
Certificates
Search by Lot NumberMETT51343101_TDS_EN
Fiche technique
Mettler InLab® Sensors
Documentation produit
Specifications
- Connection:MultiPin™
- Ø×L:12×120 mm
- For:Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samples
- Type:InLab® Expert NTC30
- Description:InLab® Expert NTC30
- Material:PEEK
- Seq:02
- Electrolyte:XEROLYT® polymer
- Temp.:0...+100 °C
- pH:0 - 14



