Orders LinkContactUsLinkComponent
Dressing jars with lid
Dressing jars with lid
180 mm, 180 mm
Catalog # BOCH8624
Supplier:   BOCHEM
Dressing jars with lid
Catalog # BOCH8624
Supplier:  BOCHEM
Dressing jars with lid
Dressing jars with lid
180 mm, 180 mm
Price
98,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Jar made of stainless steel.
  • Stackable
  • Lid with handle
  • Very good chemical and temperature resistance
Specifications
  • Ext.Ø:
    180 mm
  • Height:
    180 mm