Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dressing jars with lid

Dressing jars with lid

124 mm, 165 mm
Catalog # BOCH8623
Supplier:   BOCHEM
Dressing jars with lid
Catalog # BOCH8623
Supplier:  BOCHEM
Dressing jars with lid
Dressing jars with lid

124 mm, 165 mm

Price
58,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Jar made of stainless steel.

  • Stackable
  • Lid with handle
  • Very good chemical and temperature resistance
Specifications
  • Ext.Ø:
    124 mm
  • Height:
    165 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.