ContactUsLinkComponent
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with CH-plug. Chamberdimension (xL) 500 x 750 mm Volu me: 160 liters 1 * 1 items
Catalog # 481-2161
Supplier:  TUTTNAUER
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with CH-plug. Chamberdimension (xL) 500 x 750 mm Volu me: 160 liters 1 * 1 items
Catalog # 481-2161
Supplier:  TUTTNAUER
Frequently Bought Together