Microscope slides, UniMark®
Slides in standard packs, Rose
Catalog # 631-1207
Supplier: MARIENFELD
Jump to:
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
- Frosted end for labelling
- Soda-lime glass, Hydrolytic Class 3
- Different colours to identify preparations securely
- Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Specifications
- Colour:Rose
- Description:Slides in standard packs
- Seq:17