Orders LinkContactUsLinkComponent
VWR®, Camera Adapters
VWR®, Camera Adapters
C-mount adapter 0,4×
Catalog # 630-0867
Supplier:   VWR Collection
VWR®, Camera Adapters
Catalog # 630-0867
Supplier:  VWR Collection
VWR®, Camera Adapters
C-mount adapter 0,4×
Price
42,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Specifications
  • Description:
    C-mount adapter 0,4×
  • Seq:
    01