Orders LinkContactUsLinkComponent
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vacuum Controller V-855 for R-210/R-215
Catalog # 531-0234
Supplier:   BUCHI
Vacuum Controller, V-850/V-855
Catalog # 531-0234
Supplier:  BUCHI
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vacuum Controller V-855 for R-210/R-215
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Delivery information: Delivered with an air vent valve and pressure sensors.
Specifications
  • Description:
    Vacuum Controller V-855 for R-210/R-215
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Communication cable for R-210/R-215
    Communication cable for R-210/R-215
    181-0138
    Log in for availability
    1 * 1 ST - 34,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Woulff bottle for V-850/V-855
    Woulff bottle for V-850/V-855
    181-0141
    Log in for availability
    1 * 1 ST - 237,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Automatic distillation probe for V-855
    Automatic distillation probe for V-855
    531-0181
    Log in for availability
    1 * 1 ST - 250,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Assembly set for R-210/R-215
    Assembly set for R-210/R-215
    531-0236
    Log in for availability
    1 * 1 ST - 76,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Remote control for V-850/V-855
    Remote control for V-850/V-855
    531-0237
    Log in for availability
    1 * 1 ST - 669,00 €