Serum pipettes
Serum pipettes, graduated, 2,5 ml, 153 mm, +, Individually wrapped, 800 pieces/carton
Catalog # GREI612361
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Serum pipette 153mm sterile (25 pieces/bag) 1 * 1.000 items Catalog #GREI612362_1000
Product GREI612362_1000 is similar to GREI612361 but manufacturer and specifications may vary.
TRANSFER PIPETTE GRAD. 1ML IND/1 STER 1 * 500 items Catalog #612-6806
Product 612-6806 is similar to GREI612361 but manufacturer and specifications may vary.
TRANSFER PIPETTE GRAD. 3ML IND/1 STER 1 * 500 items Catalog #612-6804
Product 612-6804 is similar to GREI612361 but manufacturer and specifications may vary.
Product Details & Documents
These pipettes are suitable for the rapid transfer of liquids.
Specifications
- Packed:Individually wrapped, 800 pieces/carton
- Sterile:+
- Capacity:2,5 ml
- Description:Serum pipettes, graduated
- Length:153 mm
- Seq:19
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



