Orders LinkContactUsLinkComponent
Temperature stabilisation jacket
Temperature stabilisation jacket
Temperature stabilisation jacket, 577 01
Catalog # SCOI285405516
Supplier:   SI Analytics
Temperature stabilisation jacket
Catalog # SCOI285405516
Supplier:  SI Analytics
Temperature stabilisation jacket
Temperature stabilisation jacket
Temperature stabilisation jacket, 577 01
Price
404,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
  • Support plate to facilitate the use
  • Quick-action seal to simplify the changing viscometers
  • Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.
Specifications
  • Model:
    577 01
  • Description:
    Temperature stabilisation jacket
  • Seq:
    101