Orders LinkContactUsLinkComponent
Accessories
Accessories
Filter sump surface, G 7883 (inc CD), PG 8535
Catalog # MIEL5474480
Accessories
Catalog # MIEL5474480
Supplier:  
Accessories
Accessories
Filter sump surface, G 7883 (inc CD), PG 8535
Discontinued
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    Filter sump surface
  • Seq:
    16
  • For:
    G 7883 (inc CD), PG 8535