Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions in sachets

Buffer solutions in sachets

Buffer solution 10,01, 25 °C, Sachet, 30×20 ml, 30
Catalog # 662-0543
Supplier:   Mettler - Toledo
Buffer solutions in sachets
Catalog # 662-0543
Supplier:  Mettler - Toledo
CAS Number:  
Buffer solutions in sachets
Buffer solutions in sachets

Buffer solution 10,01, 25 °C, Sachet, 30×20 ml, 30

CAS Number:  
Price
68,70 €
Log in for availability
1 * 30 ST
Product Details & Documents

Solutions in sachets ensure optimum precision.

  • Advantages
  • No danger of contamination

The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH-metre that has been calibrated with a buffer associated with NIST standards.

Delivery information: The buffers in sachets are delivered with a certificate of conformity.

Specifications

Specifications

  • Pack type:
    Sachet
  • Pk:
    30
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution 10,01
  • Pk Info:
    30×20 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.