Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Bags without Closure

Sample Bags without Closure

1200×800 mm, 0,05 mm
Catalog # SCAC2078
Sample Bags without Closure
Catalog # SCAC2078
Sample Bags without Closure
Sample Bags without Closure

1200×800 mm, 0,05 mm

Price
156,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Bags made from PE transparent, for the storage or transport of samples.

Available in several sizes.

Caution: Supplied without closure ties.

Specifications
  • L×W:
    1200×800 mm
  • Seq:
    21
  • Thickness:
    0,05 mm