Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium iodide ≥99%, GPR RECTAPUR®

Potassium iodide ≥99%, GPR RECTAPUR®

Plastic bucket, 25 kg
Catalog # 26843.460
Health
Supplier:   VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Catalog # 26843.460
Supplier:  VWR Chemicals
CAS Number:  7681-11-0
Potassium iodide ≥99%, GPR RECTAPUR®
Potassium iodide ≥99%, GPR RECTAPUR®

Plastic bucket, 25 kg

CAS Number:  7681-11-0
Price
4 290,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:681 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 0.1 %
  • Fe (Iron):
    Max. 10 ppm

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.