Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KALIBRIERSTANDARD 2000 MOSM 10 X 5 ML
Catalog # ADII3LA201
Supplier:
Advanced Instruments
Certificates
Expand image
KALIBRIERSTANDARD 2000 MOSM 10 X 5 ML
Catalog # ADII3LA201
Supplier:
Advanced Instruments
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
CoC_ADII3LA201_18B0656134
Documentation
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Viscosity standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals