Screw caps, bakelite
with Butyl/PTFE seal, 48 mm, Black
Catalog # SPVAP08045
Supplier: GRAVIS
Jump to:
Product Details & Documents
Documents
SPVAP08028_TDS_EN
Datasheet
Specifications
- Thread:48 mm
- Colour:Black
- Description:with Butyl/PTFE seal
- Seq:08