ContactUsLinkComponent
Thermometer, mercury, ASTM105C, white backed, measuring range:198:252/0.2C, length:395mm, 100mm immersion, durable pigment, governm ent verified with verification certificate. 1 * 1 items
Catalog # AMARA301430CC
Supplier:  Amarell
Thermometer, mercury, ASTM105C, white backed, measuring range:198:252/0.2C, length:395mm, 100mm immersion, durable pigment, governm ent verified with verification certificate. 1 * 1 items
Catalog # AMARA301430CC
Supplier:  Amarell
Frequently Bought Together