Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, Clear, 3, 1,5 mm
Catalog # HECH42412010
Cavity slides
Catalog # HECH42412010
Cavity slides
Cavity slides
Cavity slides, Clear, 3, 1,5 mm
Price
260,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Cavity microscopy slides.
  • Ground edges
  • Made from clear glass
Suitable for staining applications in microscopy.
Specifications
  • No. of wells:
    3
  • Colour:
    Clear
  • Description:
    Cavity slides
  • Seq:
    03
  • Thickness:
    1,5 mm