PT1000 precision probes
Pointed PT1000 probe with 1500 mm red silicone cable, TPX 400-40, 3×120 mm
Catalog # EBRO1341-4168
Supplier: EBRO
Jump to:
Product Details & Documents
Specifications
- Description:Pointed PT1000 probe with 1500 mm red silicone cable, TPX 400-40
- Seq:10
- Ø×L:3×120 mm



