Orders LinkContactUsLinkComponent
Tip refill systems, Axygen®
Tip refill systems, Axygen®
1 - 200 µl, -, Transparent, 10 racks, 96 each (stack rack)
Catalog # 732-0956
Supplier:   Corning
Tip refill systems, Axygen®
Catalog # 732-0956
Supplier:  Corning
Tip refill systems, Axygen®
Tip refill systems, Axygen®
1 - 200 µl, -, Transparent, 10 racks, 96 each (stack rack)
Price
52,30 €
Log in for availability
1 * 960 ST
Product Details & Documents
Transparent PP, autoclavable.
  • Easy to load refill racks
  • Certified RNase-, DNase- and endotoxin-free
  • Universal fit tips have a bevelled reference and graduated marks
Specifications
  • Packed:
    10 racks, 96 each (stack rack)
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Seq:
    02
  • Tip colour:
    Transparent