Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Bags without Closure

Sample Bags without Closure

900×600 mm, 0,10 mm
Catalog # SCAC2077
Sample Bags without Closure
Catalog # SCAC2077
Sample Bags without Closure
Sample Bags without Closure

900×600 mm, 0,10 mm

Price
357,00 €
Connexion Pour la disponibilité
1 * 250 ST
Product Details & Documents

Bags made from PE transparent, for the storage or transport of samples.

Available in several sizes.

Caution: Supplied without closure ties.

Spécifications
  • L×W:
    900×600 mm
  • Seq:
    17
  • Thickness:
    0,10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.