Order Entry
Belgium
Orders LinkContactUsLinkComponent
Labeling tapes, warning tapes

Labeling tapes, warning tapes

Warning tape, self-adhesive PVC, 0,138 mm thick, Yellow/black, 75,0 mm
Catalog # BRDY255313
Supplier:   Brady
Labeling tapes, warning tapes
Catalog # BRDY255313
Supplier:  Brady
Labeling tapes, warning tapes
Labeling tapes, warning tapes

Warning tape, self-adhesive PVC, 0,138 mm thick, Yellow/black, 75,0 mm

Price
51,40 €
Log in for availability
1 * 1 ROU
Product Details & Documents

Self-adhesive warning film for marking pillars, pylons, the edges of walls, gate arches and lifts.

  • Extremely durable
  • Waterproof, oil, alkali and acid resistant
  • Different colours available

Length: 33,0 m.

Specifications
  • Width:
    75,0 mm
  • Colour:
    Yellow/black
  • Description:
    Warning tape, self-adhesive PVC, 0,138 mm thick
  • Seq:
    04
  • Cat. No.:
    BRDY255313