Order Entry
Belgium
Orders LinkContactUsLinkComponent
FlashGel™ DNA system

FlashGel™ DNA system

FlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tier
Catalog # 733-1615
Supplier:   LONZA
FlashGel™ DNA system
Catalog # 733-1615
Supplier:  LONZA
FlashGel™ DNA system
FlashGel™ DNA system

FlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tier

Price
225,00 €
Log in for availability
1 * 9 ST
Product Details & Documents

FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.

  • Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
  • Gel runs can be monitored at the bench, in real time without UV light
  • At least five times more sensitive than gels stained with ethidium bromide

Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel™ Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.

Delivery information: Supplied as a complete system (including FlashGel™ dock), separate components or as a starter kit (without FlashGel™ camera). The markers and dye are in a ready to load format, optimised for use with the system and recommended for best performance. The starter kit contains 9× FlashGel™ DNA cassettes (1,2%, 12+1 well); 1 ml FlashGel™ loading dye; and 150 µl FlashGel™ DNA marker (100 bp to 4 Kb).

Caution: Gel cannot be accessed. Unsuitable for recovery.

Documents

Specifications
  • Description:
    FlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tier
  • Seq:
    08
  • Cat. No.:
    733-1615
  • Separation range (bp):
    1,2% agarose: 50 - 4000
    2,2% agarose: 10 - 1000
  • Cassette size W×D×H:
    107×115×17 mm
  • Dock size W×D×H:
    120×134×54 mm
  • Gel size W×L:
    84×70 mm
  • Storage (cassettes):
    Room temperature for up to 5 months from manufacture
  • Model:
    FlashGel™ DNA
  • Well volume:
    2 - 5 µl
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Formaldehyde sample buffer
    Formaldehyde sample buffer
    733-1739
    Log in for availability
    1 * 5 mL - 84,10 €

Product Information

  • Catalog #
    Availability
    Price
  • RNA marker (0,5 - 9 kb), 50 µL
    RNA marker (0,5 - 9 kb), 50 µL
    733-1623
    Log in for availability
    1 * 50 µl - 326,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
    FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
    LONZ57064
    Log in for availability
    1 * 1 KIT - 683,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettes
    FlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettes
    733-1616
    Log in for availability
    1 * 500 µl - 297,00 €

Product Information

  • Catalog #
    Availability
    Price
  • RNA marker (0,5 bp to 9 Kb)
    RNA marker (0,5 bp to 9 Kb)
    733-1734
    Log in for availability
    1 * 50 µG - 552,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applications
    FlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applications
    733-1621
    Log in for availability
    1 * 250 µl - 297,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettes
    FlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettes
    733-1620
    Log in for availability
    1 * 500 µl - 297,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettes
    FlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettes
    733-1622
    Log in for availability
    1 * 500 µl - 297,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)
    FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)
    733-1619
    Log in for availability
    1 * 5 mL - 237,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ dock
    FlashGel™ dock
    733-1609
    Log in for availability
    1 * 1 ST - 944,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ recovery buffer, 2× 500 μl
    FlashGel™ recovery buffer, 2× 500 μl
    LONZ57060
    Log in for availability
    1 * 1.000 µl - 251,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ power supply
    FlashGel™ power supply
    LONZ57068
    Log in for availability
    1 * 1 KIT - 865,00 €

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ power supply pack, includes FlashGel™ dock
    FlashGel™ power supply pack, includes FlashGel™ dock
    LONZ57069
    Log in for availability
    1 * 1 KIT - 1 710,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.