Bellow connectors
, 29/32 NS, 114 mm, 29/32 NS
Catalog # BOHLH907-10
Supplier: Bohlender
Jump to:
Product Details & Documents
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
- Universal chemical resistance
- Suitable for vacuum applications
- Temperature resistance from −200 to +250 °C
- Compensates vibrations from vacuum pumps
- Suiltable for length compensation of heated columns and angular misalignment
Specifications
- Rodage mâle:29/32 NS
- Rodage femelle:29/32 NS
- Seq:07
- Longueur totale:114 mm



