FEETS RUBBER FOR ENVIRO-GENIE 1 * 4 items
Catalog # SCIIHWP0004
Supplier: SCIENTIFIC INDUSTRIES ELMIS
FEETS RUBBER FOR ENVIRO-GENIE 1 * 4 items
Catalog # SCIIHWP0004
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Frequently Bought Together
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Paper towel rolls, zetRoll®
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Accessories for accu-jet® pro
Supplier: Brand
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF