Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
22×60 mm
Catalog # 537-0000
Supplier:   PRAT DUMAS
Extraction thimbles
Catalog # 537-0000
Supplier:  PRAT DUMAS
Extraction thimbles
Extraction thimbles
22×60 mm
Price
114,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifications
  • Int.Ø×H:
    22×60 mm