Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
34×80 mm
N° de catalogue 537-0007
Fournisseur:   PRAT DUMAS
Extraction thimbles
N° de catalogue 537-0007
Fournisseur:  PRAT DUMAS
Extraction thimbles
Extraction thimbles
34×80 mm
Prix
122,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifications
  • Int.Ø×H:
    34×80 mm