Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
10* ml, 10/19, 0,08
Catalog # 612-3708
Supplier:   Brand
Volumetric flasks, class B
Catalog # 612-3708
Supplier:  Brand
Volumetric flasks, class B
Volumetric flasks, class B
10* ml, 10/19, 0,08
Price
10,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
High clarity PP, with PP stopper.
  • Calibrated to contain (TC, In)
  • Ring mark and inscriptions in blue
  • Exposure to temperatures up to +60 °C (autoclaving) will not cause permanently exceeded error limits, Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions
Error limits according to class B, DIN EN ISO 1042.
*Wide neck.
Specifications
  • NS:
    10/19
  • Capacity:
    10* ml
  • Tolerance:
    0,08