Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample bags, Duratec

Sample bags, Duratec

3 l, 25×25 mm
Catalog # 568-0694
Supplier:   VARIAN
Sample bags, Duratec
Catalog # 568-0694
Supplier:  VARIAN
Sample bags, Duratec
Sample bags, Duratec

3 l, 25×25 mm

Price
712,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Specifications
  • Capacity:
    3 l
  • L×W:
    25×25 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.