Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rectangular cover glasses

Rectangular cover glasses

Coverslips for Leighton tubes, 10,5×35 mm, 0,13 - 0,16 mm
Catalog # BELC1916-10535A
Marketsource
Supplier:   BELLCO GLASS
Rectangular cover glasses
Catalog # BELC1916-10535A
Supplier:  BELLCO GLASS
Marketsource
Rectangular cover glasses
Rectangular cover glasses

Coverslips for Leighton tubes, 10,5×35 mm, 0,13 - 0,16 mm

Price
35,08 €
Log in for availability
1 * 100 ST
Product Details & Documents

These rectangular cover glasses are designed for use with culture tubes.

Specifications
  • Description:
    Coverslips for Leighton tubes
  • L×W:
    10,5×35 mm
  • Seq:
    002
  • Thickness:
    0,13 - 0,16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.