Face shields
Anti-UV (UV 380), Cellulose acetate, 195×412 mm
Catalog # 111-1577
Supplier: UVEX
Jump to:
Product Details & Documents
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
- Twist ratchet enables simple and reliable adjustment to any head size
- Flip-up mechanism allows the visor to be set in any position
- Easy lens replacement
Certifications: EN 166
Documents
UVEX9708.514_TDS_EN
Datasheet
Specifications
- Type:Anti-UV (UV 380)
- L×W:195×412 mm
- Material:Cellulose acetate
Accessories
Product Information
- Catalog #MaterialAvailabilityPrice
- Replacement visor lens
Product Information
- Catalog #MaterialAvailabilityPrice
- Replacement visor lens, anti-fogReplacement visor lens, anti-fog111-8229Cellulose acetateLog in for availability1 * 1 ST - 33,00 €