Order Entry
Belgium
Orders LinkContactUsLinkComponent
Brushes for test tubes

Brushes for test tubes

16 mm, Wire, 220 mm
Catalog # MURRL340/01
Supplier:   MURRAY S
Brushes for test tubes
Catalog # MURRL340/01
Supplier:  MURRAY S
Brushes for test tubes

16 mm, Wire, 220 mm

Price
29,30 €
Log in for availability
1 * 10 ST
Product Details & Documents

Test tube brushes with bristles on wire handle.

Specifications
  • Brush head Ø:
    16 mm
  • Handle material:
    Wire
  • Total length:
    220 mm