Order Entry
Belgium
Orders LinkContactUsLinkComponent
Technical pH buffer solutions, HI 5000 series

Technical pH buffer solutions, HI 5000 series

1,00, ±0,01, 25 °C, 500 mL
Catalog # HANNHI5001
Supplier:   Hanna
Technical pH buffer solutions, HI 5000 series
Catalog # HANNHI5001
Supplier:  Hanna
CAS Number:  
Technical pH buffer solutions, HI 5000 series

1,00, ±0,01, 25 °C, 500 mL

CAS Number:  
Price
43,30 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • Colour coded labels
  • ±0,01 pH

 : Technical buffer solutions with certificate.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,01
  • pH value:
    1,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.