Orders LinkContactUsLinkComponent
Dressing jars
Dressing jars
200 mm, 200 mm
Catalog # MURRG500/08
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG500/08
Supplier:  MURRAY S
Dressing jars
200 mm, 200 mm
Price
94,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Ext.Ø:
    200 mm
  • Height:
    200 mm
  • Seq:
    07