Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Dressing jars

Dressing jars

100 mm, 200 mm
Catalog # MURRG500/03
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG500/03
Supplier:  MURRAY S
Dressing jars

100 mm, 200 mm

Price
43,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dressing jars, made of moulded glass, round.

  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Ø ext.:
    100 mm
  • Hauteur:
    200 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.