Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test tubes, without rim, light wall

Test tubes, without rim, light wall

75 mm, 10 mm, 0,8 mm
Catalog # MURRG021/31
Supplier:   MURRAY S
Test tubes, without rim, light wall
Catalog # MURRG021/31
Supplier:  MURRAY S
Test tubes, without rim, light wall

75 mm, 10 mm, 0,8 mm

Price
39,20 €
Log in for availability
1 * 100 ST
Product Details & Documents

Glass, clear.

  • Excellent chemical resistance
  • Good temperature resistance
Specifications
  • Ext.Ø:
    10 mm
  • Wall thickness:
    0,8 mm
  • Length:
    75 mm