Dissection sets
Product Details & Documents
These high quality dissecting instruments are designed for everyday use. They are subject to stringent manufacturing quality procedures, and therefore suitable for surgical applications.
The Basic Set includes:
Two scalpels
Two needles
One seeker
One blunt forceps
One sharp forceps
One fine scissors
One blunt scissors
The Advanced Set includes:
Three scalpels
Two needles
One seeker
One section lifter
One blunt forceps
One sharp forceps
One sharp curved forceps
One fine scissors
One blunt scissors
One section razor
Specifications
- Description:Dissection kit, Basic Set in PVC roll, 9 pcs.
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.