Orders LinkContactUsLinkComponent
Dissecting scissors, curved tip
Dissecting scissors, curved tip
Curved, sharp, fine point, 105 mm
Catalog # 233-0159
Supplier:   MURRAY S
Dissecting scissors, curved tip
Catalog # 233-0159
Supplier:  MURRAY S
Dissecting scissors, curved tip
Dissecting scissors, curved tip
Curved, sharp, fine point, 105 mm
Price
5,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Closed or open shanks
  • Curved, sharp/sharp
Specifications
  • Type:
    Curved, sharp, fine point
  • Length:
    105 mm