Orders LinkContactUsLinkComponent
Accessories for data loggers, Dickson®
Accessories for data loggers, Dickson®
Software and serial cable for data loggers, DicksonWare™ s
Catalog # DICKA015
Supplier:   DICKSON
Accessories for data loggers, Dickson®
Catalog # DICKA015
Supplier:  DICKSON
Accessories for data loggers, Dickson®
Software and serial cable for data loggers, DicksonWare™ s
Price
169,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Specifications
  • Description:
    Software and serial cable for data loggers, DicksonWare™ s