Pipette tips
Standard, 1000 - 5000 µl, -, Transparent, Finnpipette®, Treff, Brand, Rack
KART9847
: KARTELL
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- Tips in racks come with snap-on lid
- Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
- Containers and tips are autoclavable to +121 °C for 20 minutes
- Packed:Rack
- Sterile:-
- Volume:1000 - 5000 µl
- Seq:50
- Tip style:Standard
- Tip colour:Transparent
- For use with:Finnpipette®, Treff, Brand
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.