Orders LinkContactUsLinkComponent
Tip boxes, dispolab®
Tip boxes, dispolab®
Box for refill filter tips in rack with transparent snap-on lid, Grey
Catalog # KART9756
Supplier:   KARTELL
Tip boxes, dispolab®
Catalog # KART9756
Supplier:  KARTELL
Tip boxes, dispolab®
Tip boxes, dispolab®
Box for refill filter tips in rack with transparent snap-on lid, Grey
Price
81,50 €
Log in for availability
1 * 10 ST
Product Details & Documents
Empty boxes for use with refill tips.
  • Semitransparent boxes in PC have a horizontal sliding cover
  • Yellow boxes in PP feature a snap-on cover
Specifications
  • Colour:
    Grey
  • Description:
    Box for refill filter tips in rack with transparent snap-on lid
  • Seq:
    03