pH buffer solutions, NIST traceable, Orion™
pH buffer in 60 ml bottle, 9,18, Colourless, Plastic bottle, 5
Perform accurate, reproducible pH calibrations with Orion™ pH buffers.
- NIST traceability
- Colour-coded solutions are available for ease of use and identification
Solutions are manufactured under ISO 9000 quality standards.
The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.
: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.
- Pack type:Plastic bottle
- Pk:5
- Colour:Colourless
- Description:pH buffer in 60 ml bottle
- pH value:9,18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



