pH buffer solutions, NIST traceable, Orion™
pH buffer in 60 ml bottle, 1,68, Colourless, Plastic bottle, 5
Catalog # ORIO9116860
Supplier: Thermo Orion
pH buffer solutions, NIST traceable, Orion™
Catalog # ORIO9116860
Supplier: Thermo Orion
CAS Number:
Jump to:
Product Details & Documents
Perform accurate, reproducible pH calibrations with Orion™ pH buffers.
- NIST traceability
- Colour-coded solutions are available for ease of use and identification
Solutions are manufactured under ISO 9000 quality standards.
The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.
Livraison: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:5
- Colour:Colourless
- Description:pH buffer in 60 ml bottle
- pH value:1,68
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



